Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGFBR3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195259.1 | NP_001182188.1 | 1 | 146 | ||
XM_011527610.2 | XP_011525912.1 | 1 | 1082 | |||
XM_011527611.2 | XP_011525913.1 | 1 | 1082 | |||
XM_011527612.2 | XP_011525914.1 | 1 | 1082 | |||
XM_011527613.2 | XP_011525915.1 | 1 | 1082 | |||
XM_011527614.2 | XP_011525916.1 | 1 | 1082 | |||
GenBank mRNA | DR033794.1 | - | 1 | 159 |