Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARFRP1 SIRNA Assays ›
Gene Symbol:
ARFRP1
Gene Name:
ADP ribosylation factor related protein 1
Gene Aliases:
ARL18, ARP, Arp1
Chromosome Location:
Chr.20: 63698642 - 63708013 on Build GRCh38
UniGene ID:
Hs.389277
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s19738
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARFRP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134758.3
NP_001128230.1
4
403
NM_001267544.2
NP_001254473.1
4
408
NM_001267545.2
NP_001254474.1
4
403
NM_001267546.2
NP_001254475.1
3
309
NM_001267547.2
NP_001254476.1
4
408
NM_001267548.2
NP_001254477.1
4
305
NM_001267549.2
NP_001254478.1
4
408
NM_003224.5
NP_003215.1
4
403
NR_051954.2
4
403
NR_051955.2
4
408
NR_051956.2
4
408
NR_051957.2
4
403
NR_051958.2
4
403
XM_011528482.2
XP_011526784.1
4
340
XM_011528483.1
XP_011526785.1
4
470
XM_017027576.1
XP_016883065.1
4
284
GenBank mRNA
AK095962.1
-
3
822
AL522492.3
-
3
260
AL533893.3
-
4
393
BC021023.1
-
4
288
BC092480.1
-
4
289
30531333
BC093967.1
-
4
306
7939812
BC143447.1
-
4
306
9051955
BC143448.1
-
4
311
9051956
BC143467.1
-
4
311
9051975
BC143477.1
-
4
311
BI524032.1
-
4
402
BI819763.1
-
4
277
BM546222.1
-
328
BM549361.1
-
371
BT019624.1
-
3
255
BT019663.1
-
3
255
BU526941.1
-
4
345
BX342690.2
-
4
289
BX342805.2
-
4
382
BX463457.2
-
4
305
CA489361.1
-
3
247
CR456752.1
-
3
255
X91504.1
-
3
266
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_029702, NM_001165995, NM_001165992, NM_001165991, CA539216, and others.
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
small GTPase mediated signal transduction
gastrulation
protein localization to Golgi apparatus
retrograde transport, endosome to Golgi
Golgi to plasma membrane protein transport
GTPase activity
protein binding
GTP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-8tsm6:80/100.66.77.47:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline