Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001711.5 | NP_001702.1 | 8 | 1182 | ||
XM_017029724.1 | XP_016885213.1 | 7 | 961 | |||
GenBank mRNA | AK092954.1 | - | 8 | 961 | ||
AK093023.1 | - | 8 | 1016 | |||
AK094059.1 | - | 9 | 991 | |||
AK222484.1 | - | 8 | 1091 | |||
AK223302.1 | - | 8 | 1062 | |||
AK291955.1 | - | 8 | 1093 | |||
AK293268.1 | - | 6 | 662 | |||
AK293289.1 | - | 8 | 1017 | |||
AK297966.1 | - | 8 | 878 | |||
AK298756.1 | - | 5 | 513 | |||
BC002416.2 | - | 8 | 1075 | 3162633 | ||
BC004244.1 | - | 8 | 1085 | 3503374 | ||
BT007323.1 | - | 7 | 946 | |||
J04599.1 | - | 8 | 1066 |