Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277285.1 | NP_001264214.1 | 19 | 4047 | ||
XM_006718795.3 | XP_006718858.1 | 8 | 2561 | |||
XM_011542691.2 | XP_011540993.1 | 20 | 4734 | |||
XM_011542692.2 | XP_011540994.1 | 20 | 4722 | |||
XM_011542694.2 | XP_011540996.1 | 20 | 4737 | |||
XM_011542695.2 | XP_011540997.1 | 19 | 4265 | |||
XM_017017393.1 | XP_016872882.1 | 19 | 4752 | |||
XM_017017395.1 | XP_016872884.1 | 21 | 4782 | |||
XM_017017396.1 | XP_016872885.1 | 13 | 4076 | |||
GenBank mRNA | BG009128.1 | - | 1 | 175 |