Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005752.4 | NP_005743.4 | 2 | 251 | ||
GenBank mRNA | AK313316.1 | - | 2 | 251 | ||
AY358376.1 | - | 2 | 203 | |||
BC104414.2 | - | 2 | 240 | 40003144 | ||
BC104415.2 | - | 2 | 240 | 40003149 | ||
DA998903.1 | - | 2 | 251 |