Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRTM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134745.1 | NP_001128217.1 | 4 | 2048 | ||
NM_001282924.1 | NP_001269853.1 | 4 | 1978 | |||
XM_011533117.2 | XP_011531419.1 | 3 | 1954 | |||
GenBank mRNA | AK122612.1 | - | 4 | 1970 | ||
AY358324.1 | - | 4 | 1957 |