Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUGP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017392.4 | NP_001017392.2 | 7 | 2630 | ||
NM_001321697.1 | NP_001308626.1 | 7 | 2786 | |||
NM_001321698.1 | NP_001308627.1 | 7 | 2578 | |||
NM_001321699.1 | NP_001308628.1 | 7 | 2578 | |||
NM_014884.4 | NP_055699.2 | 7 | 2630 | |||
XM_017026137.1 | XP_016881626.1 | 7 | 2786 | |||
GenBank mRNA | AB002363.2 | - | 7 | 2782 | ||
AF518874.1 | - | 7 | 2619 | |||
AK074842.1 | - | 8 | 2656 | |||
AK291275.1 | - | 7 | 2597 | |||
AK299860.1 | - | 7 | 2045 | |||
AL832488.1 | - | 7 | 2550 | |||
BC020586.1 | - | 7 | 2550 | 4400185 | ||
BX647813.1 | - | 7 | 2559 |