Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC728554 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003615.2 | 2 | 690 | |||
GenBank mRNA | AK310274.1 | - | 2 | 164 |
Gene Symbol: THOC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032361.3 | NP_115737.1 | 2 | 464 | ||
XM_017009985.1 | XP_016865474.1 | 3 | 331 | |||
GenBank mRNA | BG502664.1 | - | 2 | 185 | ||
BU781626.1 | - | 1 | 61 |