Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058238.2 | NP_478679.1 | 4 | 1179 | ||
XM_011530366.1 | XP_011528668.1 | 4 | 944 | |||
GenBank mRNA | AB062766.1 | - | 4 | 810 | ||
AF416743.1 | - | 4 | 1179 | |||
AK123019.1 | - | 4 | 774 | |||
AK289526.1 | - | 4 | 944 | |||
BC034923.1 | - | 4 | 902 | 5457045 |