Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306070.1 | NP_001292999.1 | 8 | 1902 | ||
NM_144778.3 | NP_659002.1 | 9 | 1990 | |||
NM_207304.2 | NP_997187.1 | 7 | 1859 | |||
XM_005254018.2 | XP_005254075.1 | 10 | 2329 | |||
XM_005254019.2 | XP_005254076.1 | 8 | 2198 | |||
XM_005254021.2 | XP_005254078.1 | 9 | 2293 | |||
XM_005254022.2 | XP_005254079.1 | 8 | 2239 | |||
XM_006719912.2 | XP_006719975.1 | 9 | 2234 | |||
XM_011521045.2 | XP_011519347.1 | 9 | 1841 | |||
XM_017020308.1 | XP_016875797.1 | 10 | 1936 | |||
XM_017020309.1 | XP_016875798.1 | 8 | 1771 | |||
XM_017020310.1 | XP_016875799.1 | 9 | 1828 | |||
XM_017020311.1 | XP_016875800.1 | 7 | 1735 | |||
GenBank mRNA | AF061261.1 | - | 11 | 1949 | ||
AF116683.2 | - | 2 | 228 | |||
AF491866.1 | - | 9 | 1931 | |||
AK291727.1 | - | 7 | 1806 | |||
AK304699.1 | - | 4 | 586 | |||
AK315844.1 | - | 7 | 1782 | |||
AY101770.1 | - | 6 | 1094 | |||
BC104038.1 | - | 7 | 1787 | 40026585 | ||
BC104039.1 | - | 8 | 1823 | 40026586 | ||
BC104040.1 | - | 9 | 1918 | 40026588 | ||
CR749802.1 | - | 8 | 1818 |