Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXNC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005761.2 | NP_005752.1 | 24 | 4150 | ||
NR_037687.1 | 5 | 694 | ||||
XM_006719186.3 | XP_006719249.1 | 23 | 4138 | |||
XM_017018672.1 | XP_016874161.1 | 24 | 4177 | |||
GenBank mRNA | AB208934.1 | - | 23 | 2831 | ||
AF030339.1 | - | 24 | 4150 | |||
AK295226.1 | - | 9 | 1279 | |||
AK304327.1 | - | 5 | 694 | |||
AK307920.1 | - | 5 | 597 | |||
AK308985.1 | - | 10 | 1083 |