Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2IRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173537.3 | NP_775808.3 | 5 | 694 | ||
GenBank mRNA | AK095257.1 | - | 2 | 253 | ||
AK225845.1 | - | 5 | 651 | |||
AK291431.1 | - | 5 | 663 | |||
AY260739.1 | - | 5 | 647 | |||
AY312853.1 | - | 5 | 647 | |||
AY312854.1 | - | 4 | 487 | |||
AY336979.1 | - | 5 | 654 | |||
AY336980.1 | - | 5 | 647 | |||
BC047706.1 | - | 5 | 647 |
Gene Symbol: GTF2IRD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003795.2 | NP_001003795.1 | 5 | 694 | ||
GenBank mRNA | AK095572.1 | - | 8, 9 | 2439 | ||
AK098269.1 | - | 5 | 679 | |||
AY312850.1 | - | 5 | 669 | |||
AY312851.1 | - | 5 | 638 | |||
BC067859.1 | - | 5 | 659 | 30340785 | ||
BC136849.1 | - | 5 | 654 | 9020840 |
Gene Symbol: GTF2IRD2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY312852.1 | - | 2 | 241 |