Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320198.1 | NP_001307127.1 | 7 | 983 | ||
XM_005268866.4 | XP_005268923.1 | 7 | 5219 | |||
XM_017019296.1 | XP_016874785.1 | 6 | 1068 | |||
GenBank mRNA | AK292237.1 | - | 7 | 954 | ||
AK297794.1 | - | 6 | 908 | |||
BC069585.1 | - | 5 | 824 | 7262333 | ||
X99142.1 | - | 5 | 766 |
Gene Symbol: KRT81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002281.3 | NP_002272.2 | 5 | 825 | ||
GenBank mRNA | BC117465.1 | - | 6 | 796 | 40126016 | |
BC117467.1 | - | 6 | 797 | 40126018 | ||
X80197.1 | - | 1 | 33 | |||
X81420.1 | - | 6 | 537 |
Gene Symbol: KRT83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002282.3 | NP_002273.3 | 5 | 853 | ||
GenBank mRNA | AK314904.1 | - | 5 | 853 | ||
BC069546.1 | - | 5 | 840 | 7262357 | ||
BC126935.1 | - | 5 | 812 | 40018026 | ||
BC128063.1 | - | 5 | 812 | 40006347 | ||
X99141.1 | - | 842 |