Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100996724 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003064.1 | XP_016858553.1 | 40 | 6513 | ||
GenBank mRNA | CR749273.1 | - | 21 | 3963 |
Gene Symbol: LOC728989 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024442.2 | 3 | 298 | |||
GenBank mRNA | AK299984.1 | - | 3 | 298 |