Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BHMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001713.2 | NP_001704.2 | 2 | 172 | ||
GenBank mRNA | AK303780.1 | - | 2 | 144 | ||
AK315224.1 | - | 2 | 144 | |||
BC012616.1 | - | 2 | 148 | 4045543 | ||
DA370221.1 | - | 2 | 173 | |||
EU794592.1 | - | 2 | 172 | |||
U50929.1 | - | 2 | 93 |