Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM86B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006716256.3 | XP_006716319.1 | 4, 5 | 375 | ||
XM_006716257.3 | XP_006716320.1 | 3, 4 | 303 | |||
XM_011543843.2 | XP_011542145.1 | 3, 4 | 251 | |||
GenBank mRNA | AK308439.1 | - | 4, 5 | 361 |
Gene Symbol: FAM86B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137610.1 | NP_001131082.1 | 4, 5 | 337 | ||
XM_005273605.4 | XP_005273662.1 | 2, 3 | 130 | |||
XM_006716387.3 | XP_006716450.1 | 3, 4 | 331 | |||
XM_011544618.2 | XP_011542920.1 | 4, 5 | 382 | |||
XM_011544619.2 | XP_011542921.1 | 3, 4 | 349 | |||
XM_011544622.2 | XP_011542924.1 | 3, 4 | 251 | |||
GenBank mRNA | AK309984.1 | - | 4, 5 | 390 |