Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166395.1 | NP_001159867.1 | 2 | 467 | ||
NM_005769.2 | NP_005760.1 | 2 | 415 | |||
GenBank mRNA | AF131235.1 | - | 2 | 385 | ||
AF149783.1 | - | 1 | 296 | |||
AF280088.1 | - | 2 | 325 | |||
AK026635.1 | - | 2 | 275 | |||
AK074746.1 | - | 2 | 272 | |||
AL709927.1 | - | 2 | 297 | |||
BC035282.1 | - | 2 | 281 | 5179826 | ||
DA045171.1 | - | 2 | 467 | |||
DA496702.1 | - | 2 | 428 |