Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK298486.1 | - | 2 | 211 | ||
AK309342.1 | - | 2 | 301 | |||
BC032550.1 | - | 2 | 209 | |||
X14362.1 | - | 2 | 107 | |||
Y00816.1 | - | 207 |