Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291815.1 | NP_001278744.1 | 88 | 13502 | ||
XM_011518465.2 | XP_011516767.1 | 88 | 13639 | |||
XM_011518466.2 | XP_011516768.1 | 88 | 13639 | |||
XM_011518467.2 | XP_011516769.1 | 87 | 13462 | |||
XM_011518468.2 | XP_011516770.1 | 88 | 13639 | |||
XM_017014585.1 | XP_016870074.1 | 67 | 10308 | |||
XM_017014586.1 | XP_016870075.1 | 42 | 6236 |