Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286699.1 | NP_001273628.1 | 2 | 457 | ||
NM_001286700.1 | NP_001273629.1 | 3 | 529 | |||
NM_005772.4 | NP_005763.3 | 4 | 705 | |||
XM_006716715.3 | XP_006716778.1 | 4 | 652 | |||
GenBank mRNA | AF067172.1 | - | 4 | 424 | ||
AF161456.1 | - | 3 | 314 | |||
AJ276894.1 | - | 4 | 447 | |||
AK022904.1 | - | 4 | 602 | |||
AK225872.1 | - | 4 | 616 | |||
AL518076.3 | - | 2 | 329 | |||
AL560629.3 | - | 3 | 396 | |||
BC001025.2 | - | 4 | 598 | 3343468 |