Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA2P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027001.1 | 3 | 566 | |||
GenBank mRNA | AK307326.1 | - | 4 | 623 | ||
AK309056.1 | - | 4 | 490 | |||
AK311265.1 | - | 3 | 488 | |||
BC054509.1 | - | 3 | 383 | |||
BC071951.1 | - | 3 | 523 | |||
BC096759.1 | - | 3 | 523 |
Gene Symbol: GOLGA2P10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026811.3 | 4 | 672 | |||
NR_033936.3 | 3 | 583 | ||||
NR_103496.2 | 3 | 533 | ||||
GenBank mRNA | AK309732.1 | - | 3 | 534 | ||
AK310769.1 | - | 3 | 486 | |||
AK311170.1 | - | 3 | 493 | |||
BC007811.2 | - | 4 | 596 | |||
BC065260.1 | - | 4 | 589 | |||
BC072390.1 | - | 3 | 536 |