Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139173.3 | NP_631912.2 | 12 | 1635 | ||
XM_006714093.3 | XP_006714156.1 | 13 | 1654 | |||
XM_011531622.1 | XP_011529924.1 | 13 | 1618 | |||
XM_011531623.1 | XP_011529925.1 | 12 | 1622 | |||
GenBank mRNA | AF447585.1 | - | 12 | 1612 | ||
BC110794.1 | - | 12 | 1519 | 5745227 | ||
BC132712.1 | - | 12 | 1562 | 40146734 | ||
BC136966.1 | - | 12 | 1562 | 9020958 | ||
BC144443.1 | - | 9 | 1100 | |||
DQ003059.1 | - | 12 | 1615 |