Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005772.4 | NP_005763.3 | 3 | 511 | ||
XM_006716715.3 | XP_006716778.1 | 3 | 458 | |||
GenBank mRNA | AF067172.1 | - | 3 | 230 | ||
AF161456.1 | - | 2 | 120 | |||
AJ276894.1 | - | 3 | 253 | |||
AK022904.1 | - | 3 | 408 | |||
AK225872.1 | - | 3 | 422 | |||
BC001025.2 | - | 3 | 404 | 3343468 | ||
DB024289.1 | - | 3 | 457 |