Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SORBS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018003.2 | NP_001018003.1 | 7 | 722 | ||
NM_005775.4 | NP_005766.3 | 18 | 1805 | |||
XM_005273371.1 | XP_005273428.1 | 9 | 950 | |||
XM_006716266.1 | XP_006716329.1 | 19 | 1768 | |||
XM_006716268.2 | XP_006716331.1 | 18 | 2169 | |||
XM_017012944.1 | XP_016868433.1 | 8 | 903 | |||
XM_017012945.1 | XP_016868434.1 | 7 | 735 | |||
XM_017012946.1 | XP_016868435.1 | 7 | 720 | |||
GenBank mRNA | AF037261.1 | - | 18 | 1606 | ||
AF064807.1 | - | 7 | 710 | |||
AK301196.1 | - | 16 | 1996 | |||
BC004314.1 | - | 3 | 257 | |||
BC010146.2 | - | 7 | 686 | |||
BC067260.1 | - | 18 | 1591 | 6186182 | ||
BC091514.1 | - | 18 | 1805 | 6201273 |