Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SORBS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018003.2 | NP_001018003.1 | 6 | 578 | ||
NM_005775.4 | NP_005766.3 | 17 | 1661 | |||
XM_005273371.1 | XP_005273428.1 | 8 | 806 | |||
XM_006716266.1 | XP_006716329.1 | 18 | 1624 | |||
XM_006716268.2 | XP_006716331.1 | 17 | 2025 | |||
XM_017012944.1 | XP_016868433.1 | 7 | 759 | |||
XM_017012945.1 | XP_016868434.1 | 6 | 591 | |||
XM_017012946.1 | XP_016868435.1 | 6 | 576 | |||
GenBank mRNA | AF037261.1 | - | 17 | 1462 | ||
AF064807.1 | - | 6 | 566 | |||
AK301196.1 | - | 15 | 1852 | |||
BC004314.1 | - | 2 | 113 | |||
BC010146.2 | - | 6 | 542 | |||
BC067260.1 | - | 17 | 1447 | 6186182 | ||
BC091514.1 | - | 17 | 1661 | 6201273 | ||
CX783871.1 | - | 6 | 578 |