Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNIH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005776.2 | NP_005767.1 | 4 | 403 | ||
GenBank mRNA | AF070654.1 | - | 4 | 356 | ||
AF104398.1 | - | 4 | 356 | |||
AK074781.1 | - | 4 | 308 | |||
AK311897.1 | - | 4 | 316 | |||
AY358635.1 | - | 4 | 313 | |||
BC103741.1 | - | 4 | 404 | 6025795 | ||
BG503618.1 | - | 3 | 205 | |||
BQ233425.1 | - | 403 | ||||
BQ937490.1 | - | 4 | 428 |