Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNIH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005776.2 | NP_005767.1 | 2 | 219 | ||
GenBank mRNA | AF070654.1 | - | 2 | 172 | ||
AF104398.1 | - | 2 | 172 | |||
AK074781.1 | - | 2 | 124 | |||
AK307074.1 | - | 2 | 124 | |||
AK311897.1 | - | 2 | 132 | |||
AY358635.1 | - | 2 | 129 | |||
BC103741.1 | - | 2 | 220 | 6025795 | ||
BG503618.1 | - | 2 | 134 | |||
BQ233425.1 | - | 219 | ||||
BQ937490.1 | - | 2 | 244 | |||
BX248284.1 | - | 2 | 112 |