Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CT47A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080145.1 | NP_001073614.1 | 3 | 1227 |
Gene Symbol: CT47A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080146.1 | NP_001073615.1 | 3 | 1227 |
Gene Symbol: CT47A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080137.1 | NP_001073606.1 | 3 | 1227 |
Gene Symbol: CT47A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173571.2 | NP_775842.2 | 3 | 1227 | ||
GenBank mRNA | BC029540.1 | - | 3 | 1231 | 5297051 |
Gene Symbol: CT47A12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242922.1 | NP_001229851.1 | 3 | 1227 |
Gene Symbol: CT47A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080144.1 | NP_001073613.1 | 3 | 1227 |
Gene Symbol: CT47A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080143.1 | NP_001073612.1 | 3 | 1227 |
Gene Symbol: CT47A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080142.1 | NP_001073611.1 | 3 | 1227 |
Gene Symbol: CT47A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080141.1 | NP_001073610.1 | 3 | 1227 |
Gene Symbol: CT47A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080140.1 | NP_001073609.1 | 3 | 1227 |
Gene Symbol: CT47A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080139.1 | NP_001073608.1 | 3 | 1227 |
Gene Symbol: CT47A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080138.1 | NP_001073607.1 | 3 | 1227 |
Gene Symbol: CT47B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145718.1 | NP_001139190.1 | 3 | 1260 | ||
XM_017029734.1 | XP_016885223.1 | 3 | 1219 |