Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005778.3 | NP_005769.1 | 10 | 947 | ||
NR_036627.2 | 9 | 873 | ||||
XM_006712917.1 | XP_006712980.1 | 10 | 947 | |||
XM_011533262.1 | XP_011531564.1 | 10 | 947 | |||
XM_017005503.1 | XP_016860992.1 | 10 | 929 | |||
XM_017005505.1 | XP_016860994.1 | 9 | 840 | |||
XM_017005506.1 | XP_016860995.1 | 7 | 690 | |||
XM_017005507.1 | XP_016860996.1 | 6 | 3721 | |||
GenBank mRNA | AB208813.1 | - | 9 | 3363 | ||
AF091263.1 | - | 10 | 920 | |||
AF103802.4 | - | 10 | 898 | |||
AK297249.1 | - | 9 | 827 | |||
AK297445.1 | - | 10 | 802 | |||
AK302766.1 | - | 9 | 755 | |||
AK314032.1 | - | 10 | 902 | |||
AU129811.1 | - | 3 | 214 | |||
BC002957.1 | - | 10 | 893 | |||
DA776815.1 | - | 3 | 168 | |||
KU178505.1 | - | 9 | 772 | |||
KU178506.1 | - | 8 | 698 | |||
U23946.1 | - | 10 | 841 |