Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHFPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005779.2 | NP_005770.1 | 4 | 722 | ||
XM_006714515.1 | XP_006714578.1 | 3 | 409 | |||
GenBank mRNA | AK125227.1 | - | 1 | 219 | ||
AY309920.1 | - | 1 | 121 | |||
BC136335.1 | - | 1 | 73 | 9020322 | ||
BC136336.1 | - | 1 | 73 | 9020323 | ||
CN314535.1 | - | 4 | 546 | |||
CR936660.1 | - | 2 | 359 |