Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000987.3 | NP_000978.1 | 3 | 311 | ||
NM_001315530.1 | NP_001302459.1 | 3 | 604 | |||
NM_001315531.1 | NP_001302460.1 | 3 | 523 | |||
GenBank mRNA | AK311804.1 | - | 3 | 257 | ||
BC071664.1 | - | 3 | 246 | 5189436 | ||
BI091167.1 | - | 3 | 311 | |||
BI596427.1 | - | 3 | 528 | |||
BI603704.1 | - | 3 | 608 | |||
BM928617.1 | - | 2 | 238 | |||
CR456828.1 | - | 2 | 215 |
Gene Symbol: MSS51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR936824.1 | - | 6 | 3828 |