Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005783.3 | NP_005774.2 | 2 | 250 | ||
XM_017003147.1 | XP_016858636.1 | 2 | 209 | |||
GenBank mRNA | AB006679.1 | - | 2 | 188 | ||
AB451403.1 | - | 1 | 59 | |||
AK222616.1 | - | 2 | 176 | |||
AK301722.1 | - | 3 | 220 | |||
AK313777.1 | - | 2 | 170 | |||
BC005968.1 | - | 2 | 186 | 4073821 | ||
BC022864.1 | - | 2 | 190 | 4096526 | ||
BC024223.2 | - | 2 | 190 | 4107372 | ||
BC070183.1 | - | 2 | 184 | 4551680 | ||
BG772935.1 | - | 2 | 317 | |||
CR456935.1 | - | 1 | 59 |