Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242826.1 | NP_001229755.1 | 4 | 536 | ||
NM_005785.3 | NP_005776.1 | 4 | 597 | |||
NM_194358.2 | NP_919339.1 | 5 | 740 | |||
NM_194359.2 | NP_919340.1 | 4 | 492 | |||
NR_040053.1 | 5 | 740 | ||||
XM_005268561.4 | XP_005268618.1 | 5 | 648 | |||
XM_011537734.2 | XP_011536036.1 | 5 | 654 | |||
XM_011537735.2 | XP_011536037.1 | 5 | 571 | |||
GenBank mRNA | AF077599.1 | - | 3 | 430 | ||
AK300752.1 | - | 5 | 598 | |||
AK304576.1 | - | 5 | 437 | |||
AK314811.1 | - | 4 | 456 | |||
BC024284.1 | - | 5 | 718 | |||
BC032637.1 | - | 3 | 445 | 5496849 | ||
BC036267.1 | - | 4 | 541 | |||
BC095418.1 | - | 5 | 675 | |||
BP201682.1 | - | 4 | 492 | |||
BU170070.1 | - | 4 | 473 | |||
CB989647.1 | - | 4 | 624 | |||
DC316022.1 | - | 4 | 536 |