Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPT7P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027269.1 | 1, 2 | 295 | |||
GenBank mRNA | BC042387.1 | - | 1, 2 | 295 | ||
BC048143.1 | - | 1, 2 | 295 | |||
BC171738.1 | - | 1, 2 | 261 |
Gene Symbol: LOC100287497 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136590.1 | 1, 2 | 329 | |||
NR_136591.1 | 1, 2 | 329 | ||||
NR_136592.1 | 3, 4 | 630 | ||||
NR_136593.1 | 1, 2 | 329 | ||||
GenBank mRNA | AK124970.1 | - | 1 | 1627 | ||
BX281887.1 | - | 2, 3 | 250 | |||
DA132394.1 | - | 1, 2 | 312 | |||
DA229030.1 | - | 1, 2 | 312 |