Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPANXA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013453.3 | NP_038481.2 | 2 | 334 | ||
GenBank mRNA | AF098306.1 | - | 2 | 310 | ||
AJ238277.1 | - | 2 | 313 | |||
AK225988.1 | - | 2 | 333 | |||
AY920732.1 | - | 2 | 274 | |||
AY920733.1 | - | 2 | 274 | |||
AY920735.1 | - | 2 | 274 | |||
AY920736.1 | - | 2 | 274 | |||
AY920737.1 | - | 2 | 274 | |||
AY920738.1 | - | 2 | 274 | |||
AY920739.1 | - | 2 | 274 | |||
AY920741.1 | - | 2 | 274 | |||
AY920742.1 | - | 2 | 274 | |||
AY920743.1 | - | 2 | 274 | |||
AY920744.1 | - | 2 | 274 | |||
AY920745.1 | - | 2 | 274 | |||
AY920746.1 | - | 2 | 274 | |||
BC054023.1 | - | 2 | 335 | 6648369 | ||
BC069393.1 | - | 2 | 310 | 7262682 | ||
BC069816.1 | - | 2 | 310 | 7262670 | ||
CB115288.1 | - | 2 | 334 |
Gene Symbol: SPANXA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145662.3 | NP_663695.1 | 2 | 334 | ||
GenBank mRNA | CB110020.1 | - | 2 | 334 | ||
CB113891.1 | - | 2 | 334 |
Gene Symbol: SPANXC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022661.3 | NP_073152.2 | 2 | 367 | ||
GenBank mRNA | AA412605.1 | - | 2 | 330 |
Gene Symbol: SPANXD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032417.3 | NP_115793.1 | 2 | 608 | ||
GenBank mRNA | AF312765.1 | - | 2 | 284 | ||
AJ457795.1 | - | 2 | 310 | |||
AY920734.1 | - | 2 | 274 | |||
AY920740.1 | - | 2 | 274 | |||
BC005382.1 | - | 2 | 367 | 3935644 | ||
BC062672.1 | - | 2 | 548 | 4047937 | ||
BC103961.1 | - | 2 | 307 | 40013988 | ||
BC121070.2 | - | 2 | 317 | 40119569 | ||
BC134418.1 | - | 2 | 317 | 40119568 | ||
BM554791.1 | - | 2 | 608 |