Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPHOSPH10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005791.2 | NP_005782.1 | 2 | 874 | ||
GenBank mRNA | AK056862.1 | - | 2 | 874 | ||
AK310931.1 | - | 2 | 556 | |||
BC041084.1 | - | 2 | 524 | |||
BC126389.1 | - | 2 | 811 | 8992105 | ||
X98494.1 | - | 2 | 497 |
Gene Symbol: LOC643802 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207030.1 | NP_001193959.1 | 1 | 242 | ||
XM_017023559.1 | XP_016879048.1 | 1 | 325 | |||
XM_017023560.1 | XP_016879049.1 | 1 | 325 | |||
GenBank mRNA | AV757420.1 | - | 1 | 312 | ||
BX537874.1 | - | 1 | 83 |