Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDM15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104260.1 | 24, 25 | 4559 | |||
NR_135464.1 | 24, 25 | 4700 | ||||
GenBank mRNA | AF426259.1 | - | 4, 5 | 2152 | ||
AF426260.1 | - | 5, 6 | 1859 | |||
AL109788.1 | - | 1, 2 | 1444 | |||
AY063456.1 | - | 6, 7 | 2175 | |||
BC067102.1 | - | 24, 25 | 4559 | 5925140 |