Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005794.3 | NP_005785.1 | 6 | 964 | ||
NM_182908.4 | NP_878912.1 | 6 | 964 | |||
XM_005267249.1 | XP_005267306.1 | 6 | 676 | |||
XM_006720001.3 | XP_006720064.1 | 6 | 639 | |||
XM_011536338.1 | XP_011534640.1 | 5 | 605 | |||
XM_011536339.2 | XP_011534641.1 | 5 | 455 | |||
GenBank mRNA | AB209653.1 | - | 4 | 4606 | ||
AK000345.1 | - | 1 | 1095 | |||
AK092834.1 | - | 2 | 2375 | |||
AK309945.1 | - | 6 | 2451 | |||
BC002786.2 | - | 6 | 955 | 3614599 | ||
BC007339.2 | - | 5 | 490 | |||
U31875.1 | - | 6 | 950 |