Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCEB3CL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100817.1 | NP_001094287.1 | 2 | 1159 | ||
GenBank mRNA | BC103912.1 | - | 2 | 1155 | ||
BC103913.1 | - | 2 | 1160 | 40006027 |
Gene Symbol: LOC107983955 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017026108.1 | XP_016881597.1 | 1 | 733 |
Gene Symbol: TCEB3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145653.3 | NP_663628.2 | 1 | 1395 | ||
GenBank mRNA | AB076840.1 | - | 1 | 1159 |
Gene Symbol: TCEB3CL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242907.1 | NP_001229836.1 | 1 | 1395 |