Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005794.3 | NP_005785.1 | 8 | 1157 | ||
NM_182908.4 | NP_878912.1 | 8 | 1168 | |||
XM_005267249.1 | XP_005267306.1 | 8 | 869 | |||
XM_006720001.3 | XP_006720064.1 | 8 | 832 | |||
XM_011536338.1 | XP_011534640.1 | 7 | 798 | |||
XM_011536339.2 | XP_011534641.1 | 7 | 648 | |||
GenBank mRNA | AB209653.1 | - | 6 | 4799 | ||
AK000345.1 | - | 1 | 1817 | |||
AK092834.1 | - | 4 | 2568 | |||
AK222857.1 | - | 8 | 1157 | |||
AK309945.1 | - | 8 | 2644 | |||
BC002786.2 | - | 8 | 1159 | 3614599 | ||
BC007339.2 | - | 7 | 683 | |||
U31875.1 | - | 8 | 1143 |