Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUTF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322038.1 | NP_001308967.1 | 5, 6 | 644 | ||
NM_001322039.1 | NP_001308968.1 | 5, 6 | 733 | |||
NM_001322040.1 | NP_001308969.1 | 4, 5 | 643 | |||
NM_001322041.1 | NP_001308970.1 | 4, 5 | 753 | |||
NM_005796.2 | NP_005787.1 | 4, 5 | 554 | |||
GenBank mRNA | AK311822.1 | - | 4, 5 | 373 | ||
BC002348.2 | - | 4, 5 | 365 | 2819267 | ||
BP216441.1 | - | 5, 6 | 530 | |||
BX394845.2 | - | 4, 5 | 656 | |||
CR456937.1 | - | 3, 4 | 271 | |||
U43939.1 | - | 3, 4 | 271 | |||
X07315.1 | - | 4, 5 | 370 |