Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005801.3 | NP_005792.1 | 2 | 322 | ||
GenBank mRNA | AF083441.1 | - | 2 | 305 | ||
AF100737.1 | - | 2 | 290 | |||
AJ012375.1 | - | 2 | 289 | |||
AK311862.1 | - | 2 | 311 | |||
AL050005.1 | - | 1 | 1023 | |||
BC005118.1 | - | 2 | 302 | 2823520 | ||
BC008710.1 | - | 2 | 296 | 3350981 | ||
BT006645.1 | - | 2 | 158 | |||
BU165055.1 | - | 2 | 339 | |||
CR457051.1 | - | 2 | 158 | |||
L26247.1 | - | 2 | 297 |