Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLOT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318875.1 | NP_001305804.1 | 8, 9 | 1101 | ||
NM_005803.3 | NP_005794.1 | 9, 10 | 1245 | |||
XM_005248780.3 | XP_005248837.1 | 9, 10 | 978 | |||
XM_006714947.3 | XP_006715010.1 | 8, 9 | 1082 | |||
XM_017010157.1 | XP_016865646.1 | 9, 10 | 899 | |||
XM_017010158.1 | XP_016865647.1 | 7, 8 | 800 | |||
GenBank mRNA | AF085357.1 | - | 9, 10 | 1034 | ||
AF089750.2 | - | 9, 10 | 1070 | |||
AK000816.1 | - | 9, 10 | 1081 | |||
AK222530.1 | - | 9, 10 | 1081 | |||
AK301291.1 | - | 8, 9 | 935 | |||
BC001146.1 | - | 8, 9 | 928 | 3534771 | ||
BC128154.1 | - | 2, 3 | 338 | |||
BT007077.1 | - | 8, 9 | 906 | |||
CR456946.1 | - | 8, 9 | 906 |