Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX39A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005804.3 | NP_005795.2 | 10 | 1333 | ||
NR_046366.2 | 9 | 1223 | ||||
XM_006722606.3 | XP_006722669.1 | 11 | 1458 | |||
XM_011527620.1 | XP_011525922.1 | 10 | 1352 | |||
XM_011527621.2 | XP_011525923.1 | 10 | 1277 | |||
GenBank mRNA | AK301847.1 | - | 11 | 1416 | ||
AL512698.1 | - | 3 | 1627 | |||
BC001009.2 | - | 10 | 1270 | 2820926 | ||
BC032128.2 | - | 9 | 1169 | 5017293 | ||
BM792110.1 | - | 4 | 466 |