Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUEDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271875.1 | NP_001258804.1 | 2 | 245 | ||
NM_001292025.1 | NP_001278954.1 | 2 | 550 | |||
XM_006721900.2 | XP_006721963.1 | 2 | 860 | |||
XM_017024640.1 | XP_016880129.1 | 2 | 215 | |||
XM_017024641.1 | XP_016880130.1 | 2 | 213 | |||
GenBank mRNA | AK000746.1 | - | 2 | 550 | ||
BC056882.1 | - | 2 | 245 | 6150503 |