Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGPTL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021146.3 | NP_066969.1 | 5 | 1278 | ||
GenBank mRNA | AB593026.1 | - | 5 | 1212 | ||
AB593027.1 | - | 5 | 1279 | |||
AK313716.1 | - | 5 | 1244 | |||
AY358301.1 | - | 5 | 1201 | |||
BC001881.1 | - | 5 | 1226 | 3544149 | ||
BT009802.1 | - | 5 | 987 | |||
Y16132.1 | - | 5 | 994 |