Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGPTL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021146.3 | NP_066969.1 | 3 | 814 | ||
XM_017000004.1 | XP_016855493.1 | 3 | 779 | |||
GenBank mRNA | AB593026.1 | - | 3 | 748 | ||
AB593027.1 | - | 3 | 815 | |||
AK313716.1 | - | 3 | 780 | |||
AY358301.1 | - | 3 | 737 | |||
BC001881.1 | - | 3 | 762 | 3544149 | ||
BT009802.1 | - | 3 | 523 | |||
Y16132.1 | - | 3 | 530 |