Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005816.4 | NP_005807.1 | 8, 9 | 1344 | ||
NM_198196.2 | NP_937839.1 | 9, 10 | 1392 | |||
NR_134917.1 | 8, 9 | 1344 | ||||
XM_006713469.3 | XP_006713532.1 | 9, 10 | 1353 | |||
XM_006713470.3 | XP_006713533.1 | 8, 9 | 1305 | |||
XM_017005521.1 | XP_016861010.1 | 8, 9 | 1164 | |||
XM_017005522.1 | XP_016861011.1 | 6, 7 | 1041 | |||
GenBank mRNA | AK290719.1 | - | 8, 9 | 1230 | ||
AK315237.1 | - | 8, 9 | 1226 | |||
AL833681.1 | - | 9, 10 | 1239 |