Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318889.1 | NP_001305818.1 | 4 | 884 | ||
NM_005816.4 | NP_005807.1 | 4 | 884 | |||
NM_198196.2 | NP_937839.1 | 5 | 932 | |||
NR_134917.1 | 4 | 884 | ||||
XM_005247063.3 | XP_005247120.1 | 5 | 893 | |||
XM_006713469.3 | XP_006713532.1 | 5 | 893 | |||
XM_006713470.3 | XP_006713533.1 | 4 | 845 | |||
XM_017005521.1 | XP_016861010.1 | 5 | 893 | |||
GenBank mRNA | AK290719.1 | - | 4 | 770 | ||
AK315237.1 | - | 4 | 766 | |||
AL833681.1 | - | 5 | 779 | |||
BC020749.1 | - | 4 | 815 | 4669495 |