Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135217.1 | NP_001128689.1 | 4, 5 | 876 | ||
NM_006992.3 | NP_008923.1 | 4, 5 | 704 | |||
NM_201650.2 | NP_964013.1 | 4, 5 | 704 | |||
GenBank mRNA | AK223535.1 | - | 4, 5 | 638 | ||
AK292291.1 | - | 4, 5 | 639 | |||
AK292367.1 | - | 4, 5 | 704 | |||
BC029858.1 | - | 4, 5 | 677 | 5170643 | ||
U72508.1 | - | 4, 5 | 596 |